Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080588.1 | NP_001074057.1 | 8 - 9 | 2156 | 65 |
XM_009302237.3 | XP_009300512.1 | 8 - 9 | 3033 | 65 | |
XM_009302238.3 | XP_009300513.1 | 8 - 9 | 3048 | 65 | |
GenBank mRNA | BC129037.1 | - | 8 - 9 | 2156 | 65 |