Assay Design



Gene Transcripts

Gene Symbol
hadhab
Entrez Gene ID
793834
Gene Name
hydroxyacyl-CoA dehydrogenase trifunctional multienzyme complex subunit alpha b
Gene Aliases
im:7144234, zgc:158138
Chromosome Location
Chr.17: 37227961 - 37252810 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089437.1 NP_001082906.1 18 - 19 2009 75
GenBank mRNA BC096816.1 - 18 - 19 75
BC131856.1 - 18 - 19 2009 75

View Full Product Details