Assay Design



Gene Transcripts

Gene Symbol
slc9a6a
Entrez Gene ID
792983
Gene Name
solute carrier family 9 member A6a
Gene Aliases
fv47b07, wu:fv47b07, zgc:163055
Chromosome Location
Chr.14: 175220 - 209508 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098256.2 NP_001091726.2 8 - 9 1227 92
XM_009291141.3 XP_009289416.1 9 - 10 1372 92

View Full Product Details