Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099254.1 | NP_001092724.1 | 8 - 9 | 1014 | 81 |
XM_005161327.4 | XP_005161384.1 | 8 - 9 | 1330 | 81 | |
XM_009295459.3 | XP_009293734.1 | 9 - 10 | 1349 | 81 | |
XM_009295460.3 | XP_009293735.1 | 8 - 9 | 1343 | 81 | |
GenBank mRNA | BC139893.1 | - | 8 - 9 | 1014 | 81 |
FJ593490.1 | - | 8 - 9 | 1026 | 81 | |
FJ593491.1 | - | 9 - 10 | 1032 | 81 |