Assay Design



Gene Transcripts

Gene Symbol
chkb
Entrez Gene ID
563589
Gene Name
choline kinase beta
Gene Aliases
fc21g05, si:ch211-218c17.2, wu:fc21g05
Chromosome Location
Chr.18: 8320156 - 8337571 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100012.2 NP_001093482.1 6 - 7 931 56
XM_005159014.3 XP_005159071.1 6 - 7 1200 56
GenBank mRNA BC163504.1 - 6 - 7 1092 56
BC163518.1 - 6 - 7 1092 56

View Full Product Details