Assay Design



Gene Transcripts

Gene Symbol
pes
Entrez Gene ID
30228
Gene Name
pescadillo
Gene Aliases
-
Chromosome Location
Chr.5: 3604 - 23843 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131030.3 NP_571105.3 13 - 14 1635 71
GenBank mRNA BC049339.1 - 13 - 14 71
U77627.1 - 13 - 14 71

View Full Product Details