Assay Design



Gene Transcripts

Gene Symbol
pes
Entrez Gene ID
30228
Gene Name
pescadillo
Gene Aliases
-
Chromosome Location
Chr.5: 3604 - 23843 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131030.3 NP_571105.3 5 - 6 666 121
GenBank mRNA BC049339.1 - 5 - 6 647 121
U77627.1 - 5 - 6 632 121

View Full Product Details