Assay Design
Gene Transcripts
- Gene Symbol
- btr30
- Entrez Gene ID
-
393394
- Gene Name
- bloodthirsty-related gene family, member 30
- Gene Aliases
-
zgc:64214
- Chromosome Location
-
Chr.19: 48139755 - 48145313 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200422.1 |
NP_956716.1 |
5 - 6 |
1175 |
62 |
GenBank mRNA |
BC054643.1 |
- |
5 - 6 |
1175 |
62 |
- Gene Symbol
- LOC110437745
- Entrez Gene ID
-
110437745
- Gene Name
- tripartite motif-containing protein 7-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 48004528 - 48010490 on Build GRCz11
View Full Product Details