Assay Design



Gene Transcripts

Gene Symbol
elovl5
Entrez Gene ID
393425
Gene Name
ELOVL fatty acid elongase 5
Gene Aliases
zgc:63549
Chromosome Location
Chr.13: 2394197 - 2417402 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200453.2 NP_956747.1 4 - 5 463 83
XM_005156490.4 XP_005156547.1 4 - 5 876 83
GenBank mRNA AF532782.2 - 4 - 5 480 83
BC055137.1 - 4 - 5 463 83
EH446014.1 - 4 - 5 462 83

View Full Product Details