Assay Design



Gene Transcripts

Gene Symbol
arl3l1
Entrez Gene ID
393666
Gene Name
ADP ribosylation factor like GTPase 3, like 1
Gene Aliases
zgc:73049
Chromosome Location
Chr.21: 33172400 - 33185577 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200693.1 NP_956987.1 5 - 6 639 67
GenBank mRNA BC059431.1 - 5 - 6 639 67
BC075770.1 - 5 - 6 671 67

View Full Product Details