Assay Design



Gene Transcripts

Gene Symbol
ptf1a
Entrez Gene ID
368662
Gene Name
pancreas associated transcription factor 1a
Gene Aliases
PTF1, si:zc142h2.2, zgc:112216
Chromosome Location
Chr.2: 29710770 - 29712071 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207641.2 NP_997524.1 1 - 2 679 79
GenBank mRNA AY245546.1 - 1 - 2 641 79
BC093269.1 - 1 - 2 679 79

View Full Product Details