Assay Design
Gene Transcripts
- Gene Symbol
- ubxn1
- Entrez Gene ID
-
322073
- Gene Name
- UBX domain protein 1
- Gene Aliases
-
fb49a05, wu:fb49a05, zgc:77531
- Chromosome Location
-
Chr.14: 45471709 - 45482615 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_212607.1 |
NP_997772.1 |
3 - 4 |
137 |
94 |
GenBank mRNA |
BC067166.1 |
- |
3 - 4 |
137 |
94 |
View Full Product Details