Assay Design



Gene Transcripts

Gene Symbol
tm9sf1
Entrez Gene ID
445156
Gene Name
transmembrane 9 superfamily member 1
Gene Aliases
fa03b09, wu:fa03b09, zgc:100810
Chromosome Location
Chr.24: 11888526 - 11908351 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003550.2 NP_001003550.1 7 - 8 1320 73
XM_021470138.1 XP_021325813.1 7 - 8 1577 73
GenBank mRNA BC078291.1 - 7 - 8 1324 73

View Full Product Details