Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006039.2 | NP_001006039.2 | 5 - 6 | 626 | 112 |
XM_005169164.4 | XP_005169221.1 | 13 - 14 | 1576 | 112 | |
XM_009305561.3 | XP_009303836.1 | 13 - 14 | 1573 | 112 | |
GenBank mRNA | BC083374.1 | - | 5 - 6 | 599 | 112 |