Assay Design



Gene Transcripts

Gene Symbol
med10
Entrez Gene ID
550394
Gene Name
mediator complex subunit 10
Gene Aliases
png, zgc:112222
Chromosome Location
Chr.19: 27970362 - 27974152 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017699.1 NP_001017699.1 3 - 4 424 142
GenBank mRNA AY698065.1 - 3 - 4 424 142
BC093274.1 - 3 - 4 424 142

View Full Product Details