Assay Design



Gene Transcripts

Gene Symbol
olah
Entrez Gene ID
568096
Gene Name
oleoyl-ACP hydrolase
Gene Aliases
zgc:136926
Chromosome Location
Chr.19: 10649425 - 10656733 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039932.1 NP_001035021.1 4 - 5 678 121
XM_005159455.3 XP_005159512.1 4 - 5 460 121
XM_021468123.1 XP_021323798.1 4 - 5 682 121
GenBank mRNA BC114310.1 - 4 - 5 678 121

View Full Product Details