Assay Design



Gene Transcripts

Gene Symbol
hsd17b2
Entrez Gene ID
449537
Gene Name
hydroxysteroid (17-beta) dehydrogenase 2
Gene Aliases
-
Chromosome Location
Chr.25: 35942867 - 35950740 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040188.1 NP_001035278.1 1 - 2 192 75
GenBank mRNA BC090448.1 - 1 - 2 130 75
DQ364628.1 - 1 - 2 192 75

View Full Product Details