Assay Design



Gene Transcripts

Gene Symbol
drc3
Entrez Gene ID
777619
Gene Name
dynein regulatory complex subunit 3
Gene Aliases
lrrc48, zgc:153749
Chromosome Location
Chr.3: 62126972 - 62139434 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077727.1 NP_001071195.1 5 - 6 637 109
XM_021471579.1 XP_021327254.1 5 - 6 646 109
GenBank mRNA BC125885.1 - 5 - 6 637 109

View Full Product Details