Assay Design



Gene Transcripts

Gene Symbol
kirrel3l
Entrez Gene ID
557315
Gene Name
kirre like nephrin family adhesion molecule 3, like
Gene Aliases
cb33, kirrel3, sb:cb33, si:dkey-29g23.1, wu:fb74e02
Chromosome Location
Chr.15: 36941455 - 37027289 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099344.2 NP_001092814.2 14 - 15 2406 74
GenBank mRNA CU638818.1 - 14 - 15 1819 74
EF571006.1 - 14 - 15 2406 74

View Full Product Details