Assay Design



Gene Transcripts

Gene Symbol
exoc3l1
Entrez Gene ID
567832
Gene Name
exocyst complex component 3-like 1
Gene Aliases
exoc3l, si:ch211-51l3.1
Chromosome Location
Chr.25: 14425836 - 14434262 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099422.1 NP_001092892.1 10 - 11 1950 139
GenBank mRNA BC139871.1 - 10 - 11 1950 139

View Full Product Details