Assay Design



Gene Transcripts

Gene Symbol
exoc3l1
Entrez Gene ID
567832
Gene Name
exocyst complex component 3-like 1
Gene Aliases
exoc3l, si:ch211-51l3.1
Chromosome Location
Chr.25: 14425836 - 14434262 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099422.1 NP_001092892.1 5 - 6 1223 126
XM_005174338.4 XP_005174395.1 5 - 6 1983 126
GenBank mRNA BC139871.1 - 5 - 6 1223 126

View Full Product Details