Assay Design



Gene Transcripts

Gene Symbol
dad1
Entrez Gene ID
566527
Gene Name
defender against cell death 1
Gene Aliases
si:ch211-9d9.5
Chromosome Location
Chr.2: 51640849 - 51644034 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100439.1 NP_001093909.1 1 - 2 270 114
GenBank mRNA BC150399.1 - 1 - 2 270 114

View Full Product Details