Assay Design



Gene Transcripts

Gene Symbol
crybb1l3
Entrez Gene ID
566891
Gene Name
crystallin, beta B1, like 3
Gene Aliases
zgc:171636
Chromosome Location
Chr.15: 28096152 - 28104403 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102390.1 NP_001095860.1 2 - 3 145 79
GenBank mRNA BC150363.1 - 2 - 3 145 79

View Full Product Details