Assay Design



Gene Transcripts

Gene Symbol
dla
Entrez Gene ID
30131
Gene Name
deltaA
Gene Aliases
cb335, fa04c10, wu:fa04c10
Chromosome Location
Chr.1: 54013455 - 54026180 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130954.2 NP_571029.2 8 - 9 1660 102
GenBank mRNA AF030031.1 - 8 - 9 1651 102
BC075742.1 - 8 - 9 1660 102
CU458989.1 - 8 - 9 1651 102
DQ993185.1 - 8 - 9 1651 102

View Full Product Details