Assay Design



Gene Transcripts

Gene Symbol
atoh7
Entrez Gene ID
58216
Gene Name
atonal bHLH transcription factor 7
Gene Aliases
ath5, zath5
Chromosome Location
Chr.13: 25449699 - 25450724 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131632.1 NP_571707.1 1 - 1 355 61
GenBank mRNA AB049457.1 - 1 - 1 355 61
BC071520.1 - 1 - 1 365 61

View Full Product Details