Assay Design



Gene Transcripts

Gene Symbol
mxe
Entrez Gene ID
337663
Gene Name
myxovirus (influenza virus) resistance E
Gene Aliases
wu:fj93f01, zgc:111895
Chromosome Location
Chr.9: 32824572 - 32839888 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182867.1 NP_878287.1 6 - 7 929 125
XM_005167664.4 XP_005167721.2 7 - 8 1061 125
GenBank mRNA AJ544827.1 - 6 - 7 929 125
BC095587.1 - 7 - 8 971 125

View Full Product Details