Assay Design



Gene Transcripts

Gene Symbol
mcm4
Entrez Gene ID
337598
Gene Name
minichromosome maintenance complex component 4
Gene Aliases
cb1025, fc12c09, fj85g09, hm:zeh1616, wu:fc12c09, wu:fj85g09, zeh1616
Chromosome Location
Chr.24: 35536662 - 35561672 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198913.2 NP_944595.2 6 - 7 589 68
GenBank mRNA BC056514.1 - 6 - 7 583 68
BC065958.1 - 6 - 7 589 68

View Full Product Details