Assay Design



Gene Transcripts

Gene Symbol
slc6a19b
Entrez Gene ID
326030
Gene Name
solute carrier family 6 member 19b
Gene Aliases
slc6a19, wu:fd58f10, zgc:66050
Chromosome Location
Chr.16: 18974074 - 18998549 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199736.1 NP_956030.1 10 - 11 1562 94
GenBank mRNA BC059804.1 - 10 - 11 1562 94
BC154319.1 - 10 - 11 1564 94

View Full Product Details