Assay Design
Gene Transcripts
- Gene Symbol
- slc6a19b
- Entrez Gene ID
-
326030
- Gene Name
- solute carrier family 6 member 19b
- Gene Aliases
-
slc6a19, wu:fd58f10, zgc:66050
- Chromosome Location
-
Chr.16: 18974074 - 18998549 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199736.1 |
NP_956030.1 |
11 - 12 |
1726 |
120 |
GenBank mRNA |
BC059804.1 |
- |
11 - 12 |
1726 |
120 |
View Full Product Details