Assay Design



Gene Transcripts

Gene Symbol
map1lc3c
Entrez Gene ID
393268
Gene Name
microtubule-associated protein 1 light chain 3 gamma
Gene Aliases
KM0005, zgc:56565
Chromosome Location
Chr.11: 44798247 - 44801898 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200298.2 NP_956592.1 3 - 4 233 90
GenBank mRNA AB264295.1 - 3 - 4 216 90
AY398373.1 - 3 - 4 233 90
BC049489.1 - 3 - 4 248 90

View Full Product Details