Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_200768.1 | NP_957062.1 | 4 - 5 | 303 | 96 |
XM_005158766.4 | XP_005158823.1 | 5 - 6 | 410 | 96 | |
XM_005158767.4 | XP_005158824.1 | 4 - 5 | 319 | 96 | |
XM_009293125.3 | XP_009291400.1 | 5 - 6 | 410 | 96 | |
GenBank mRNA | BC059599.1 | - | 4 - 5 | 303 | 96 |