Assay Design



Gene Transcripts

Gene Symbol
gar1
Entrez Gene ID
393950
Gene Name
GAR1 homolog, ribonucleoprotein
Gene Aliases
nola1, zgc:56086
Chromosome Location
Chr.14: 232316 - 243831 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200975.2 NP_957269.2 3 - 4 447 117
GenBank mRNA BC045900.1 - 3 - 4 420 117
EH495404.1 - 3 - 4 476 117

View Full Product Details