Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213092.1 | NP_998257.1 | 10 - 11 | 1166 | 129 |
XM_005167390.4 | XP_005167447.1 | 10 - 11 | 1256 | 129 | |
GenBank mRNA | BC044401.1 | - | 10 - 11 | 1186 | 129 |
BC066527.1 | - | 10 - 11 | 1166 | 129 | |
BC091971.1 | - | 10 - 11 | 1188 | 129 |