Assay Design
Gene Transcripts
- Gene Symbol
- adgrl4
- Entrez Gene ID
-
406676
- Gene Name
- adhesion G protein-coupled receptor L4
- Gene Aliases
-
eltd1, fd10a02, wu:fd10a02, zgc:63629
- Chromosome Location
-
Chr.11: 7383213 - 7410314 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_213367.2 |
NP_998532.2 |
11 - 12 |
1628 |
75 |
GenBank mRNA |
BC055171.1 |
- |
11 - 12 |
1625 |
75 |
View Full Product Details