Assay Design
Gene Transcripts
- Gene Symbol
- srsf6a
- Entrez Gene ID
-
406775
- Gene Name
- serine and arginine rich splicing factor 6a
- Gene Aliases
-
fa12h12, sfrs6a, wu:fa12h12, wu:fa13e06, zgc:63770
- Chromosome Location
-
Chr.23: 4260437 - 4271984 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_213466.1 |
NP_998631.1 |
2 - 3 |
222 |
94 |
GenBank mRNA |
BC055238.1 |
- |
2 - 3 |
222 |
94 |
View Full Product Details