Assay Design



Gene Transcripts

Gene Symbol
fga
Entrez Gene ID
378986
Gene Name
fibrinogen alpha chain
Gene Aliases
sb:cb892, wu:fa55c12, wu:fb20e08, wu:fb59b03, wu:fi39d04, zgc:92117
Chromosome Location
Chr.1: 10066100 - 10071522 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194989.1 NP_001181918.1 5 - 6 1486 61
GenBank mRNA BC075895.1 - 5 - 6 1425 61

View Full Product Details