Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002153.3 | NP_001002153.2 | 1 - 2 | 299 | 63 |
NM_001167855.2 | NP_001161327.1 | 2 - 3 | 210 | 63 | |
GenBank mRNA | BC071358.1 | - | 1 - 2 | 199 | 63 |
EH451895.1 | - | 2 - 3 | 172 | 63 |