Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012366.1 | NP_001012366.1 | 6 - 7 | 818 | 58 |
XM_005172254.4 | XP_005172311.1 | 6 - 7 | 894 | 58 | |
GenBank mRNA | AY835383.1 | - | 6 - 7 | 818 | 58 |
BC108043.1 | - | 6 - 7 | 882 | 58 |