Assay Design



Gene Transcripts

Gene Symbol
stk16
Entrez Gene ID
550323
Gene Name
serine/threonine kinase 16
Gene Aliases
zgc:112393
Chromosome Location
Chr.1: 5422437 - 5434261 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017630.1 NP_001017630.1 3 - 4 453 81
GenBank mRNA BC093283.1 - 3 - 4 453 81
BC164081.1 - 2 - 3 344 81

View Full Product Details