Assay Design



Gene Transcripts

Gene Symbol
atad1b
Entrez Gene ID
368412
Gene Name
ATPase family AAA domain containing 1b
Gene Aliases
atad1, bZ1G13.1, si:bz1g13.1, wu:fc15a10, zgc:110042
Chromosome Location
Chr.12: 17436904 - 17456083 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024421.2 NP_001019592.1 9 - 10 1141 61
GenBank mRNA BC095151.1 - 9 - 10 1142 61
BC164188.1 - 8 - 9 1007 61

View Full Product Details