Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029947.1 | NP_001025118.1 | 1 - 2 | 176 | 67 |
GenBank mRNA | AF529203.1 | - | 1 - 2 | 176 | 67 |
AM422092.1 | - | 1 - 2 | 204 | 67 | |
BC163672.1 | - | 1 - 2 | 238 | 67 |