Assay Design



Gene Transcripts

Gene Symbol
nnr
Entrez Gene ID
321023
Gene Name
nanor
Gene Aliases
-
Chromosome Location
Chr.15: 1589813 - 1590796 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029947.1 NP_001025118.1 3 - 4 387 114
GenBank mRNA AF529203.1 - 3 - 4 387 114

View Full Product Details