Assay Design



Gene Transcripts

Gene Symbol
cyp4v7
Entrez Gene ID
562008
Gene Name
cytochrome P450, family 4, subfamily V, polypeptide 7
Gene Aliases
zgc:154042
Chromosome Location
Chr.14: 290435 - 305832 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079996.1 NP_001073465.1 4 - 5 648 114
GenBank mRNA BC125940.1 - 4 - 5 648 114
BC165384.1 - 4 - 5 615 114

View Full Product Details