Assay Design



Gene Transcripts

Gene Symbol
col11a1a
Entrez Gene ID
565402
Gene Name
collagen, type XI, alpha 1a
Gene Aliases
wu:fb34e11, wu:fc11b03, zgc:158335
Chromosome Location
Chr.24: 28764019 - 28893344 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083844.1 NP_001077313.1 29 - 30 2482 94
XM_005162813.1 XP_005162870.1 30 - 31 2809 94
XM_005162814.1 XP_005162871.1 30 - 31 2806 94
XM_005162815.1 XP_005162872.1 29 - 30 2722 94
XM_005162818.1 XP_005162875.1 29 - 30 2512 94
XM_021470209.1 XP_021325884.1 28 - 29 2425 94
XM_021470210.1 XP_021325885.1 23 - 24 1589 94
GenBank mRNA BC133083.1 - 29 - 30 2482 94
GQ485664.1 - 30 - 31 2703 94

View Full Product Details