Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_131060.1 | NP_571135.1 | 12 - 13 | 1808 | 65 |
GenBank mRNA | AF086761.1 | - | 12 - 13 | 1808 | 65 |
BC162381.1 | - | 12 - 13 | 1521 | 65 | |
BC162398.1 | - | 12 - 13 | 1521 | 65 | |
CU458855.1 | - | 12 - 13 | 1451 | 65 |