Assay Design



Gene Transcripts

Gene Symbol
sema3aa
Entrez Gene ID
30266
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3Aa
Gene Aliases
sema1a, sema3a1, semaZ1a
Chromosome Location
Chr.4: 7414 - 61242 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131060.1 NP_571135.1 7 - 8 1172 85
GenBank mRNA AF086761.1 - 7 - 8 1172 85
BC162381.1 - 7 - 8 885 85
BC162398.1 - 7 - 8 885 85

View Full Product Details