Assay Design



Gene Transcripts

Gene Symbol
tpma
Entrez Gene ID
30324
Gene Name
alpha-tropomyosin
Gene Aliases
TPM1, fb37a09, tm, wu:fb37a09
Chromosome Location
Chr.7: 29952249 - 29960619 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131105.2 NP_571180.1 9 - 10 897 67
GenBank mRNA AF180892.1 - 9 - 10 908 67
BC062870.1 - 9 - 10 897 67

View Full Product Details