Assay Design



Gene Transcripts

Gene Symbol
bmp2a
Entrez Gene ID
30631
Gene Name
bone morphogenetic protein 2a
Gene Aliases
bmp2, wu:fc59d09
Chromosome Location
Chr.17: 4227274 - 4231388 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131359.1 NP_571434.1 1 - 2 321 78
GenBank mRNA BC163048.1 - 1 - 2 372 78
KM820423.1 - 1 - 2 321 78
U82233.1 - 1 - 2 321 78

View Full Product Details