Assay Design



Gene Transcripts

Gene Symbol
zdhhc8b
Entrez Gene ID
352941
Gene Name
zinc finger DHHC-type palmitoyltransferase 8b
Gene Aliases
zdhhc5, zisp
Chromosome Location
Chr.5: 17881080 - 18007077 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178304.3 NP_840089.2 4 - 5 859 60
GenBank mRNA AB085761.1 - 4 - 5 551 60
AY871211.1 - 4 - 5 859 60
BC049439.1 - 4 - 5 653 60

View Full Product Details