Assay Design
Gene Transcripts
- Gene Symbol
- klhl41b
- Entrez Gene ID
-
321064
- Gene Name
- kelch-like family member 41b
- Gene Aliases
-
cb52, kbtbd10, kbtbd10b, sarcosin, sb:cb52, wu:fb24f05, wu:fc15e07
- Chromosome Location
-
Chr.6: 3680554 - 3690209 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198979.1 |
NP_945330.1 |
4 - 5 |
1657 |
76 |
GenBank mRNA |
BC054912.1 |
- |
4 - 5 |
1657 |
76 |
View Full Product Details